Learn More
R&D Systems™ Recombinant Human IL-2R beta Fc/IL-2R gamma Fc Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3560 |
| Molecular Weight (g/mol) | M.W.-observed: 62-68 kDa (IL-2R beta) & 85-93 kDa (IL-2R gamma), M.W.-theroretical: 51 kDa (IL-2R beta) & 54 kDa (IL-2 R gamma) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human IL-2 R beta/IL-2 R gamma protein Human IL-2RB (Ala27-Asp239) Accession # (N-terminus)P14784.1 IEGR Human IgG1 (Glu99-Lys330) (with modifications) Human IL-2RG (Leu23-Asn254) Accession # (N-terminus)P31785.1 IEGR Human IgG1 (Glu99-Lys330) (with modifications) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD122, CD132, CIDX, High affinity IL-2 receptor subunit beta, IL15RB, IL-2 receptor subunit beta, IL-2RG, IMD4, IMD63, P64, SCIDX |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.