Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-3 GMP Protein, CF

Catalog No. BT003GMP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT003GMP050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BT003GMP050 Supplier R&D Systems™ Supplier No. BT003GMP050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.0150-0.150 ng/mL. The specific activity of GMP Recombinant Human IL-3 is >1.7 x 106 IU/mg, which is calibrated against the human IL-3 WHO International Standard (NIBSC code: 91/510).

Specifications

Accession Number AAC08706.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3562
Molecular Weight (g/mol) MolecularWeight-observed: 14 kDa, under reducing conditions., MolecularWeight-theroretical: 15 kDa
Quantity 50 μg
Source E. coli-derived human IL-3 protein Ala20 - Phe152, with and without an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20 °C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, ≤ -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in sterile water.
Purity or Quality Grade >97%, by SDS-PAGE with quantitative densitometry by Coomassie™ Blue Staining. The molecular weight by mass spectrometry is 15201 Da ± 50 Da, and a second 15070 Da ± 50 Da product may be present.
Protein IL-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.