Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-34 His-tag Protein, CF

Catalog No. 11449IL010
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-449-IL010 10 μg
11-449-IL01M 1 mg
11-449-IL050 50 μg
11-449-IL250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-449-IL010 Supplier R&D Systems™ Supplier No. 11449IL010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier-Free Recombinant Protein

Measured by its ability to induce CCL2/MCP-1 secretion by human peripheral blood mononuclear cells (PBMC). The ED50 for this effect is 15.0-150 ng/mL.

Specifications

Accession Number AAH29804.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 146433
Molecular Weight (g/mol) M.W. (Observed): 19-25 kDa, under reducing conditions., M.W. (Theoretical): 21 kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-34 protein Asn21-Val193, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias C16orf77, IL34, IL-34, interleukin 34
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-34
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.