Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-35 Fc Chimera Protein
GSA_VA

Catalog No. 8608IL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-08I-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 86-08I-L050 Supplier R&D Systems™ Supplier No. 8608IL050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-35 Fc Chimera Protein is derived from HEK293. The Recombinant Human IL-35 Fc Chimera Protein has been validated for the following applications: Bioactivity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 10148
Molecular Weight (g/mol) Observed M.W.: 81-97kDa, reducing conditions, Theoretical M.W.: 73kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human IL-35 protein Human IL-27 beta/EBI3 (Arg21-Lys229) Accession &Num; Q14213 (GGGS)4 Human IL-12 alpha/p35 Arg23-Ser219 Accession &Num; P29459 IEGRMD Human IgG1 Pro100-Lys330)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name IL-35
Gene Symbol EBI3
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human IL-12 R beta 2 Fc Chimera is immobilized at 5μg/mL (100μL/well), the concentration of Recombinant Human IL-35 Fc Chimera that produces 50% optimal binding response is approximately 20-120ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human IL-27 beta/EBI3 (Arg21-Lys229) Accession # Q14213, N-terminus; (GGGS)4; Human IL-12 alpha/p35 Arg23-Ser219 Accession # P29459; IEGRMD; Human IgG1 Pro100-Lys330), C-terminus
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.