Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-37/IL-1F7 Protein

Catalog No. 9225IL025
Change view
Click to view available options
Recombinant Human IL-37/IL-1F7 Protein
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
92-25I-L025 Recombinant Human IL-37/IL-1F7 Protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-25I-L025 Supplier R&D Systems™ Supplier No. 9225IL025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-37/IL-1F7 Protein is derived from E. coli. The Recombinant Human IL-37/IL-1F7 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number Q9NZH6-2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein.
Gene ID (Entrez) 27178
Molecular Weight (g/mol) 21 kDa
Name FIL1, FIL1 zeta, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, IL37, interleukin 1 family, member 7 (z
Quantity 25μg
Source E. coli-derived human IL-37/IL-1F7 protein Ser2-Asp192
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Biological Activity Measured by its ability to inhibit LPS-stimulated IL-8 secretion by HK-2 human kidney epithelial cells. The ED50 for this effect is 0.25-1.25 μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.