Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-4 Protein
GSA_VA

Catalog No. p-4720098
Change view
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
204IL010 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
204IL001MGC 1 mg Lyophilized from a 0.2μm filtered solution in PBS.
204IL010CF 10 μg Lyophilized from a 0.2μm filtered solution in PBS.
204IL050 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
204IL050CF 50 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 204IL010 Supplier R&D Systems™ Supplier No. 204IL010
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.05-0.2 ng/mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3565
Molecular Weight (g/mol) 15 kDa
Name IL-4
Quantity 10 μg
Source E. coli-derived human IL-4 protein His25-Ser153,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.