Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-6/IL-6R alpha Protein Chimera
SDP

Catalog No. 8954SR025CF
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8954SR025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8954SR025CF Supplier R&D Systems™ Supplier No. 8954SR025/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-6/IL-6R alpha Protein Chimera is derived from HEK293. The Recombinant Human IL-6/IL-6R alpha Protein Chimera has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 3570
Molecular Weight (g/mol) Observed M.W.: 78 - 93kDa, reducing conditions, Theoretical M.W.: 59kDa
Name IL-6R alpha
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human IL-6/IL-6R alpha Complex protein, (N-terminus) Human IL-6R alpha (Leu20-Asp358) Accession &Num; P08887 GGGSGGGSGGGS Human IL-6 Val30-Met212 Accession &Num; P05231 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias BSF-2, CD126, gp80, IL6, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor, MGI-2A
Common Name IL-6 R alpha
Gene Symbol IL6
Biological Activity Measured by its ability to inhibit M1 mouse myeloid leukemia cell proliferation. The ED50 for this effect is typically 2.5-12.5ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human IL-6 R alpha (Leu20-Asp358) Accession # P08887, N-terminus; GGGSGGGSGGGS; Human IL-6 Val30-Met212 Accession # P05231, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Protein IL-6/IL-6 R alpha Complex
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.