Learn More
Creative Biomart GMP Grade Recombinant Human IL21 protein
Supplier:  Creative Biomart IL21371THP
Encompass_Preferred
Recombinant Human IL21 protein GMP Grade(NP_001193935) was expressed in E. coli. Measured by its ability to induced Interferon gamma secretion by human natural killer lymphoma NK-92 cells. The ED50 for this effect is 0.4-2 ng/mL.
Catalog No. 50-208-5423