Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IMP Dehydrogenase 1/IMPDH1 Protein

Catalog No. 8904DH050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8904DH050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8904DH050 Supplier R&D Systems™ Supplier No. 8904DH050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IMP Dehydrogenase 1/IMPDH1 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IMP Dehydrogenase 1/IMPDH1 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number NP_899066
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl, EDTA, Glycerol and TCEP.
Gene ID (Entrez) 3614
Molecular Weight (g/mol) Observed M.W.: 53-64kDa, reducing conditions, Theoretical M.W.: 61kDa
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IMP Dehydrogenase 1/IMPDH1 protein Glu2-Tyr563,with a N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias DKFZp781N0678, EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 1, IMP (inosine monophosphate) dehydrogenase 1, IMP dehydrogenase 1, IMPD, IMPD 1, IMPD1, IMPDH 1, IMPDH1, IMPDH-I, inosine-5'-monophosphate dehydrogenase 1, LCA11, LCA11RP10, retinitis pigmentosa 10 (autosomal dominant), RP10, sWSS2608
Common Name IMPDH1
Gene Symbol IMPDH1
Biological Activity Measured by its ability to convert the substrate inosine-5'-phosphate (IMP) to xanthosine-5'-phosphate (XMP). The specific activity is >100 pmol/min/μg, as measured under the described conditions.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Glu2-Tyr563, with a N-terminal Met and 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.