Learn More
R&D Systems™ Recombinant Human Integrin a5ss1 Protein, CF
Description
Specifications
Specifications
| Accession Number | P08648 & P05556 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3678 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 110-140 kDa, under reducing conditions., MolecularWeight-theroretical: 108 kDa ( alpha 5 subunit), 82.3 kDa ( beta 1 subunit) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Integrin alpha 5 beta 1 protein Human Integrin alpha 5 (Phe42-Tyr995) (N-terminus) Accession # P08648 Acidic Tail Human Integrin beta 1 (Gln21-Asp728) (N-terminus) Accession # P05556 Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.20 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.