Learn More
R&DSystems™ Recombinant Human Integrin alpha 4 beta 7/LPAM 1 Protein, CF
Description
Specifications
Specifications
| Accession Number | P13612.3 (Integrin alpha 4) & P26010.1 (Integrin beta 7) |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3676 |
| Molecular Weight (g/mol) | M.W.-observed: 115-135 kDa (Integrin beta 7) & 140-160 kDa (Integrin alpha 4), under reducing conditions., M.W.-theroretical: 130 kDa (Integrin alpha 4) & 104 kDa (Integrin beta 7) |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Integrin alpha 4 beta 7/LPAM-1 protein Human ITGA4 (Tyr34-Gln970,R591L & R878Q) (N-terminus) Accession # P13612.3 IEGR Human IgG1 (Glu99-Lys330) (with modifications) Human ITGB7 (Glu20-His723) (N-terminus) Accession # P26010.1 IEGR Human IgG1 (Glu99-Lys330) (with modifications) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49d/beta7, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), LPAM1, LPAM-1, MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.