Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&DSystems™ Recombinant Human Integrin alpha 5 beta 1 Fc Protein, CF

Catalog No. 11538A5050
Change view
Click to view available options
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-538-A5050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-538-A5050 Supplier R&DSystems™ Supplier No. 11538A5050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fc Chimera

Measured by its binding ability in a functional ELISA. Recombinant Human Integrin alpha 5 beta 1 Fc Chimera (Catalog # 11538-A5) binds Human Fibronectin (Catalog # 1918-FN ) with an ED50 of 0.600-9.00 ng/mL.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3678
Molecular Weight (g/mol) M.W.-observed: 225-370 kDa, under non-reducing conditions, M.W.-theroretical: 131 kDa (Integrin alpha 4) & 105 kDa (Integrin beta 1)
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Integrin alpha 5 beta 1 protein Human ITGA5 (Phe42-Tyr995) (N-terminus) Accession # P08648.2 IEGR Human IgG1 (Pro99-Lys330) (with modifications) Human ITGB1 (Gln21-Asp728) (N-terminus) Accession # P05556.2 IEGR Human IgG1 (Pro99-Lys330) (with modifications) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Integrin alpha 5 beta 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.