Learn More
R&D Systems™ Recombinant Human Integrin alpha V beta 3 Fc Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3685 |
| Molecular Weight (g/mol) | MolecularWeight-observed: >190 kDa, under non-reducing conditions., MolecularWeight-theroretical: 133 kDa (Integrin alpha V) & 103 kDa (Integrin beta 3) |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Integrin alpha V beta 3 protein Human ITGAV (Phe31-Val992) (N-terminus) Accession # AAA36808.1 IEGR Human IgG1 (Glu99-Lys330) (with modifications) Human ITGB3 (Gly27-Asp718) (N-terminus) Accession # P05106.2 HPIEGR Human IgG1 (Glu99-Lys330) (with modifications) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.