Learn More
R&D Systems™ Recombinant Human Integrin alpha V beta 5 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3685 |
| Molecular Weight (g/mol) | 115.4 kDa ( alphaV subunit) & 84.8 kDa (beta5 subunit) |
| Name | Integrin alpha V beta 5 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha V beta 5 protein, (N-terminus) Human Integrin alpha V (Phe31-Val992) Accession &Num; P06756 His-Pro GGGSGGGS Acidic Tail 6-His tag Human Integrin beta 5 (Gly24-Asn719) Accession &Num; P18084 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.