Learn More
Biomatik Corporation Recombinant Human Interleukin-12 receptor subunit beta-2 (IL12RB2), partial
Supplier:  Biomatik Corporation RPC25507
Encompass_Preferred
Recombinant Human Interleukin-12 receptor subunit beta-2 (IL12RB2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IL12RB2. Target Synonyms: I12R2_HUMAN; IL-12 receptor subunit beta-2; IL-12R subunit beta-2; IL-12R-beta-2; IL-12RB2; IL12 receptor beta 2; IL12R beta2; IL12RB2; Interleukin 12 receptor beta 2; Interleukin 12 receptor beta 2 chain; Interleukin-12 receptor subunit beta-2; RP11 102M16.1. Accession Number: Q99665. Expression Region: 24~622aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 69.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5209