Learn More
R&D Systems™ Recombinant Human ISG15 AMC Protein
Description
Specifications
Specifications
| Accession Number | P05161.5 |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 9636 |
| Molecular Weight (g/mol) | M.W. Theroretical: 18 kDa |
| Name | ISG15/UCRP AMC |
| Quantity | 50 μg |
| Source | E. coli-derived human ISG15/UCRP protein Met1 - Gly156 (C78S, G157C) with a C-terminal 6-His tag; C-terminal AMC derivatized protein |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | 7-Amino-4-Methyl Coumarin |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.