Learn More
Creative Biomart Palifermin, Recombinant human KGF
Supplier:  Creative Biomart THP0113
Encompass_Preferred
The product is a recombinant human keratinocyte growth factor (KGF). It is 140 residues long, and is produced using E. coli.
Catalog No. 50-208-4164