Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human KGF/FGF-7 Protein, CF

Catalog No. BTKGF500
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTKGF500 500 μg
BTKGF100 100 μg
BTKGF020 20 μg
BTKGF050 50 μg
BTKGF01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. BTKGF500 Supplier R&D Systems™ Supplier No. BTKGF500
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Measured in a cell proliferation assay using Ba/F3 mouse pro B cells transfected with human FGF RIIb. The ED50 for this effect is 6.00-60.0 ng/mL.

Specifications

Accession Number P21781.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with Trehalose.
Gene ID (Entrez) 2252
Molecular Weight (g/mol) MolecularWeight-observed: 20 kDa, reducing conditions., MolecularWeight-theroretical: 19 kDa
Quantity 500 μg
Source E. coli-derived human KGF/FGF-7 protein Cys32-Thr194, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Conjugate Unconjugated
Reconstitution Reconstitute the 20 μg size at 200 μg/mL in water. Reconstitute all other sizes at 500 μg/mL in water.
Protein KGF/FGF-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.