Learn More
R&D Systems™ Recombinant Human KIR2DL1/CD158a Fc Chimera Protein
Description
Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately.
Specifications
Specifications
| Accession Number | P43626 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3802 |
| Molecular Weight (g/mol) | 51 kDa (monomer) |
| Name | KIR2DL1/CD158a Fc Chimera Protein |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human KIR2DL1/CD158a protein, (N-terminus) Human KIR2DL1/CD158a (His22-Arg242) Accession &Num; P43626 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked Homodimer |
| Conjugate | Unconjugated |
| Show More |
For Reasearch Use Only; Not for Use in Humans
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.