Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LAIR2 Fc Chimera Protein

Catalog No. 10166LR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-166-LR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-166-LR050 Supplier R&D Systems™ Supplier No. 10166LR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

LAIR2 (leukocyte-associated Ig-like receptor-2; CD306) is a secreted, 131 amino acid (aa) protein that contains one Ig-like C2 type domain, making it a member of the Ig superfamily

  • Measured by its binding ability in a functional ELISA.
  • When Bovine Collagen I is coated at 10 μg/mL, 100 μL/well, Recombinant Human LAIR2 Fc Chimera binds with an ED50 of 0.04-0.32 ng/mL.

Specifications

Accession Number Q6ISS4
For Use With (Application) Bioactivity
Gene ID (Entrez) 3904
Molecular Weight (g/mol) 41 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human LAIR2 protein

Human LAIR2 (Gln22-Pro152) Accession # Q6ISS4IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.