Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LAR Protein

Catalog No. 9377PF050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-77P-F050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-77P-F050 Supplier R&D Systems™ Supplier No. 9377PF050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human LRFN5 Fc Chimera (Catalog # 9385-SA ) is used at 1 μg/mL, the concentration of Recombinant Human LAR that produces 50% optimal binding response is 1.5-9 μg/mL.

Specifications

Accession Number P10586-2
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA and Trehalose.
Gene ID (Entrez) 5792
Molecular Weight (g/mol) M.W. (Observed): 114-146 kDa and 14 kDa, reducing conditions; M.W. (theoretical): 136 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human LAR protein Ala27-Glu1251 with a C-terminal 6-His tag, and its proteolyzed forms Ala27-Arg1169 and Gln1170-Glu1251 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias LCA-homolog, protein tyrosine phosphatase, receptor type, F, PTPRF, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LAR/PTPRF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.