Learn More
R&D Systems™ Recombinant Human Latent TGF-beta 1 Protein, CF
Description
Specifications
Specifications
| Accession Number | P01137 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 7040 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 36-42 kDa & 9-13 kDa, under reducing conditions, MolecularWeight-theroretical: 29 kDa (LAP) & 13 kDa (Mature) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human TGF-beta 1 protein Leu30-Ser390 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 2 weeks, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CEDLAP, DPD1, latency-associated peptide, TGF beta, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.