Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LH alpha/beta Heterodimer Protein

Catalog No. 8899LH010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8899LH010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8899LH010 Supplier R&D Systems™ Supplier No. 8899LH010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human LH alpha/beta Heterodimer Protein is derived from CHO. The Recombinant Human LH alpha/beta Heterodimer Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P01215 ( alpha chain) & P01229 ( beta chain)
For Use With (Application) Bioactivity
Molecular Weight (g/mol) Observed M.W.: 15-26kDa, reducing conditions, Theoretical M.W.: 10kDa ( alpha chain), 13kDa ( beta chain)
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human LH alpha/beta Heterodimer protein Ala25-Ser116 ( alpha chain) & Ser21-Leu141 ( beta chain)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name LH alpha/beta Heterodimer
Biological Activity Measured by its ability to induce cAMP accumulation in MLTC-1 mouse Leydig tumor cells. Rebois, R.V. et al. (1982) J. Cell Biol. 94:70. The ED50 for this effect is typically 0.25-2.5ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ala25-Ser116 ( alpha chain) & Ser21-Leu141 ( beta chain)
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.