Learn More
Proteintech Group Inc Recombinant Human LIF 10ug
SDP
Catalog No. 501973963 Shop All Proteintech Group Inc Products
Encompass_Preferred
Recombinant Human LIF is a pleiotropic factor produced by numerous cell types which includes myelomonocytic lineages, T cells, fibroblasts, melanoma, liver, and heart. LIF promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. Other activities include the stimulation of acute phase protein synthesis by hepatocytes, stimulation of differentiation of cholinergic nerves, and suppression of adipogenesis by inhibiting the lipoprotein lipase in adipocytes. Mature human LIF (180 aa) shares 78%, 82%, 91%, 88 and 87% aa sequence identity with mouse, rat, canine, bovine, and porcine LIF, respectively. To reduce spontaneous differentiation, LIF is typically added to stem cell culture medium.
Catalog No. 50-197-3963 Supplier Proteintech Group Inc Supplier No. HZ129210UG
Add to Cart
Add to Cart
Recombinant Human LIF is a pleiotropic factor produced by numerous cell types which includes myelomonocytic lineages, T cells, fibroblasts, melanoma, liver, and heart. LIF promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. Other activities include the stimulation of acute phase protein synthesis by hepatocytes, stimulation of differentiation of cholinergic nerves, and suppression of adipogenesis by inhibiting the lipoprotein lipase in adipocytes. Mature human LIF (180 aa) shares 78%, 82%, 91%, 88 and 87% aa sequence identity with mouse, rat, canine, bovine, and porcine LIF, respectively. To reduce spontaneous differentiation, LIF is typically added to stem cell culture medium.