Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LIF, Animal-Free Protein

Catalog No. QK0360500
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QK0360500 500 μg
QK0360050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. QK0360500 Supplier R&D Systems™ Supplier No. QK0360500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
No significant difference between EC50 of reference and test lots

Specifications

Accession Number P15018.1
For Use With (Application) Bioactivity
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 3976
Molecular Weight (g/mol) M.W. (Observed): Monomeric human LIF protein only, M.W. (Theroretical): 17 kDa
Quantity 500 μg
Source E. coli-derived human LIF protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Conjugate Unconjugated
Reconstitution Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired.
Protein LIF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.