Learn More
R&D Systems™ Recombinant Human LILRB3/CD85a/ILT5 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | AAB68668 |
| For Use With (Application) | Binding Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 11025 |
| Molecular Weight (g/mol) | M.W. (Observed): 95-105 kDa, reducing conditions; M.W. (theoretical): 72.6 kDa (monomer) |
| Name | ILT5/CD85a Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human LILRB3/CD85a/ILT5 protein Human ILRB3/CD85a/ILT5 (Gly24-Glu443) (N-terminus) Accession # AAB68668 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide linked homodimer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.