Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LILRB4/CD85k/ILT3 Fc Biotin Protein, CF

Catalog No. BT10429050
Change view
Click to view available options
:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BT10429050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT10429050 Supplier R&D Systems™ Supplier No. BT10429050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fc Chimera Biotinylated

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human LILRB4/CD85k/ILT3 Fc Chimera binds to Human Fibronectin Protein (Catalog # 1918-FN ) with an ED50 of 0.400-4.00 μg/mL.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 11006
Molecular Weight (g/mol) M.W.-observed: 50-70 kDa, under reducing conditions, M.W.-theroretical: 53 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human LILRB4/CD85k/ILT3 protein Human LILRB4 (Gly24-Glu259) (N-terminus) Accession # NP_001265355.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus )
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4
Conjugate Biotin
Reconstitution Reconstitute at 500 μg/mL in water.
Protein ILT3/CD85k/LILRB4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.