Learn More
Abcam Recombinant Human Liver Carboxylesterase 1/CES1 protein, 10UG
Catalog No. 502658050
Encompass_Preferred
Recombinant Human Liver Carboxylesterase 1/CES1 protein is a Human Fragment protein, in the 19 to 562 aa range, expressed in HEK 293, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: CES2, SES1, CES1, Liver carboxylesterase 1, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, Carboxylesterase 1, Cholesteryl ester hydrolase, Cocaine carboxylesterase, Egasyn, HMSE, Methylumbelliferyl-acetate deacetylase 1, Monocyte/macrophage serine esterase, Retinyl ester hydrolase, Serine esterase 1, Triacylglycerol hydrolase, ACAT, CE-1, hCE-1, CEH, REH, TGH
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-8050 Supplier Abcam Supplier No. AB15186410UG
Recombinant Human Liver Carboxylesterase 1/CES1 protein is a Human Fragment protein, in the 19 to 562 aa range, expressed in HEK 293, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: CES2, SES1, CES1, Liver carboxylesterase 1, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, Carboxylesterase 1, Cholesteryl ester hydrolase, Cocaine carboxylesterase, Egasyn, HMSE, Methylumbelliferyl-acetate deacetylase 1, Monocyte/macrophage serine esterase, Retinyl ester hydrolase, Serine esterase 1, Triacylglycerol hydrolase, ACAT, CE-1, hCE-1, CEH, REH, TGH
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement