Learn More
R&D Systems™ Recombinant Human Livin alpha Protein
Description
Specifications
Specifications
| Accession Number | Q96CA5 |
| For Use With (Application) | Inhibition Assay |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 79444 |
| Molecular Weight (g/mol) | M.W. (Observed): 38 kDa, reducing conditions; M.W. (theoretical): 34 kDa |
| Name | Livin α/KIAP |
| Quantity | 100 μg |
| Source | E. coli-derived human Livin protein Met1-Ser298, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Structural Form | Monomer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.