Learn More
Biomatik Corporation Recombinant Human Long-wave-sensitive opsin 1 (OPN1LW)
Supplier:  Biomatik Corporation RPC27091
Encompass_Preferred
Recombinant Human Long-wave-sensitive opsin 1 (OPN1LW) is a purified CF Transmembrane Protein. Purity: >85% as determined by SDS-PAGE. Host: In Vitro E. coli Expression System. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: OPN1LW. Target Synonyms: Long-wave-sensitive opsin 1(Red cone photoreceptor pigment; Red-sensitive opsin; ROP). Accession Number: P04000. Expression Region: 1~364aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 43.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7926