Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LRRC52 Fc Chimera Protein

Catalog No. 10260LR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-260-LR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-260-LR050 Supplier R&D Systems™ Supplier No. 10260LR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to inhibit neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human LRRC52 Fc Chimera, immobilized at 2.5 μg/mL on a 96 well plate, is able to significantly inhibit neurite outgrowth.

Specifications

Accession Number Q8N7C0
For Use With (Application) Bioactivity
Gene ID (Entrez) 440699
Molecular Weight (g/mol) 52 kDa
Name BK Channel Auxiliary Gamma Subunit LRRC52, FLJ25811, leucine rich repeat containing 52, leucine-rich repeat-containing protein 52
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human LRRC52 protein

Human LRRC-52 (Ser24-Asp244) Accession # Q8N7C0IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.