Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Lymphotoxin alpha1/beta2 Protein

Catalog No. 8884LY025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8884LY025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8884LY025 Supplier R&D Systems™ Supplier No. 8884LY025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Lymphotoxin alpha1/beta2 Protein is derived from HEK293. The Recombinant Human Lymphotoxin alpha1/beta2 Protein has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 4049
Molecular Weight (g/mol) Observed M.W.: 65-76kDa, reducing conditions, Theoretical M.W.: 61kDa
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human Lymphotoxin protein, (N-terminus) rhLT alpha (Leu35-Leu205) Accession &Num; P01374 GGGGS rhLT beta (Gln49-Gly244) Accession &Num; Q06643 GGGGS rhLT beta (Gln49-Gly244) Accession &Num; Q06643 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name Lymphotoxin
Gene Symbol LTA
Biological Activity Measured by its ability to induce IL-8 secretion by A375 human melanoma cells. The ED50 for this effect is typically 3-15ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived rhLT alpha (Leu35-Leu205) Accession # P01374, N-terminus; GGGGS; rhLT beta (Gln49-Gly244) Accession # Q06643; GGGGS; rhLT beta (Gln49-Gly244) Accession # Q06643, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.