Learn More
Biomatik Corporation Recombinant Human Lysosomal acid lipase/cholesteryl ester hydrolase (LIPA)
Supplier:  Biomatik Corporation RPC24795
Encompass_Preferred
Recombinant Human Lysosomal acid lipase/cholesteryl ester hydrolase (LIPA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LIPA. Target Synonyms: Cholesteryl esteraseLipase ASterol esterase. Accession Number: P38571. Expression Region: 22~399aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 45kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5813