Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Lysosomal alpha-Glucosidase/GAA

Catalog No. 8329GH025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-29G-H025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-29G-H025 Supplier R&D Systems™ Supplier No. 8329GH025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to release glucose from starch. The specific activity is >7,500 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P10253
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 2548
Molecular Weight (g/mol) M.W. (Observed): 95-105 kDa, reducing conditions; M.W. (theoretical): 99 kDa
Name Lysosomal alpha-Glucosidase
Quality Control Testing SDS-PAGE: 95-105kDa, reducing conditions
Quantity 25 μg
Source Human embryonic kidney cell, HEK293-derived human Lysosomal alpha-Glucosidase protein Ala70-Cys952, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 6 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Termination His
Without Additives Carrier Protein
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol GAA
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
Protein LYAG/GAA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.