Learn More
InVitria Lysobac - Recombinant Human Lysozyme - 10g
Supplier:  InVitria 777LYS01610G
Encompass
InVitria’s Lysobac recombinant human lysozyme is completely animal- and blood-free. It is commonly used as a preservative in food, beverages, diagnostic applications, bioprocessing, and life science research. Lysobac has identical amino acid sequence and physiochemical properties to the native human form of lysozyme. It exhibits four times the hydrolytic activity against gram-positive bacteria compared to commercially available chicken-egg lysozyme.Made in the USA.
Catalog No. NC1750183