Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human lysyl oxidase-like 2 (LOXL2) | >95.0% | 50 UG
SDP

Catalog No. 5000294473
Encompass_Preferred

Recombinant human lysyl oxidase-like 2 (LOXL2) expressed in CHO cells with a C-terminal His tag. The active enzyme (~85.5 kDa) is supplied lyophilized (50 μg) and formulated for stability; suitable for enzymatic activity assays and studies of extracellular matrix remodeling and EMT-related biology.

  • Expression system: CHO cells.
  • C-terminal His tag for purification and detection.
  • Supplied quantity: 50 μg, lyophilized.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Specific activity >2.0 pmol/min/μg (benzylamine assay).
  • Formulation: 20 mM MES, 50 mM NaCl, 5% trehalose, 5% mannitol, 0.01% Tween 80; store frozen.

Catalog No. 50-002-94473 Supplier Medchemexpress LLC Supplier No. HYP7477150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human lysyl oxidase-like 2 (LOXL2) expressed in CHO cells with a C-terminal His tag. The active enzyme (~85.5 kDa) is supplied lyophilized (50 μg) and formulated for stability; suitable for enzymatic activity assays and studies of extracellular matrix remodeling and EMT-related biology.

  • Expression system: CHO cells.
  • C-terminal His tag for purification and detection.
  • Supplied quantity: 50 μg, lyophilized.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Specific activity >2.0 pmol/min/μg (benzylamine assay).
  • Formulation: 20 mM MES, 50 mM NaCl, 5% trehalose, 5% mannitol, 0.01% Tween 80; store frozen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.