Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human M-CSF, Animal-Free Protein

Catalog No. p-7182944
Click to view available options
Quantity:
25 μg
5 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells

  • ED50 for this effect is 0.5 to 1.5 ng/mL.
  • The specific activity of Recombinant Human M-CSF is approximately 1.68 x 105 IU/μg, which is calibrated against recombinant human M-CSF WHO International Standard (NIBSC code: 89/512).

Specifications

Accession Number NP_757350.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5 kDa (monomer)
Name colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930
Quantity 5 μg
Source E. coli-derived human M-CSF protein Glu33-Ser190, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol CSF1
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.