Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MD-2 Protein

Catalog No. 1787MD050CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1787MD050CF Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
1787MD050 Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1787MD050CF Supplier R&D Systems™ Supplier No. 1787MD050/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human MD-2 Protein is derived from E. coli. The Recombinant Human MD-2 Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 23643
Molecular Weight (g/mol) 19.2 kDa (monomer)
Name MD-2
Quantity 50 μg
Source E. coli-derived human MD2 protein, (N-terminus) Met Human MD-2 (Glu17-Asn160) Accession &Num; BAA78717 IEGRGGGSGGGSGGGS 10-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.