Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MDL-1/CLEC5A Protein

Catalog No. 8544CL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-44C-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-44C-L050 Supplier R&D Systems™ Supplier No. 8544CL050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human MDL-1/CLEC5A Protein is derived from NS0. The Recombinant Human MDL-1/CLEC5A Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9NY25
For Use With (Application) Bioactivity
Gene ID (Entrez) 23601
Molecular Weight (g/mol) Observed M.W.: 34-45kDa, reducing conditions, Theoretical M.W.: 20kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human MDL-1/CLEC5A protein Tyr26-Lys188,with an N-terminal 9-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CLEC5A, CLECSF5, CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL1, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1
Common Name MDL-1/CLEC5A
Gene Symbol CLEC5A
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human Galectin-9 is coated at 0.5μg/mL (100μL/well), the concentration of Recombinant Human MDL-1/CLEC5A that produces 50% optimal binding response is typically 3-18ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Tyr26-Lys188, with an N-terminal 9-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.