Learn More
R&D Systems™ Recombinant Human MINDY3 Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | X mg/mL (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% (v/v) Glycerol, 2 mM TCEP |
| Gene ID (Entrez) | 80013 |
| Molecular Weight (g/mol) | M.W. (theoretical): 50 kDa |
| Quantity | 25 μg |
| Source | E. coli-derived human MINDY3 protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Species | Human |
| Buffer | X mg/mL (X μm) in 50 mm HEPES pH 7.5, 100 mm NaCl, 10% (v/v) Glycerol, 2 mm TCEP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.