Learn More
Sino Biological Recombinant Human MMP-2 Protein, Biotinylated, HPLC-verified 20 µg
Catalog No. 5000699088
Encompass_Preferred
A DNA sequence encoding the native human MMP2 (NP_004521.1) (Met 1-Cys 660) was expressed and purified. The purified protein was biotinylated in vitro.
Catalog No. 50-006-99088 Supplier Sino Biological Supplier No. 10082HNAHB20
A DNA sequence encoding the native human MMP2 (NP_004521.1) (Met 1-Cys 660) was expressed and purified. The purified protein was biotinylated in vitro.