Learn More
AnaSpec Recombinant Human MOG (1-125), Highly Purified Protein, Applications in Neuroscience Research, 5 mg
Supplier:  AnaSpec AS551581000
Encompass
Recombinant Human MOG (1-125) is a highly purified protein derived from the extracellular domain of Myelin Oligodendrocyte Glycoprotein, essential for research in neuroscience and multiple sclerosis. Its purity of ≥95% and rigorous purification through IMAC ensures reliable performance in various assays, including ELISA and Western Blotting. This product facilitates in vitro and in vivo studies, distinguishing it with low endotoxin levels and specific applicability in autoimmune research.
Catalog No. NC2313146