Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MOG Protein

Catalog No. 8535MO050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-35M-O050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-35M-O050 Supplier R&D Systems™ Supplier No. 8535MO050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human MOG Protein is derived from NS0. The Recombinant Human MOG Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q16653
For Use With (Application) Bioactivity
Gene ID (Entrez) 4340
Molecular Weight (g/mol) Observed M.W.: 20-30kDa, reducing conditions, Theoretical M.W.: 16kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human MOG protein Gly30-Gly154,with a substitution at Asp131Ala and a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein
Common Name MOG
Gene Symbol MOG
Biological Activity Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human MOG, immobilized at 1μg/mL on a 96-well plate, is able to significantly enhance neurite outgrowth.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Gly30-Gly154, with a substitution at Asp131Ala and a C-terminal 10-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.