Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat GDF-8/Myostatin Protein, CF

Catalog No. 11658G8250
Change view
Click to view available options
Quantity:
100 μg
20 μg
250 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-658-G8250 250 μg
11-658-G8050 50 μg
11-658-G8020 20 μg
11-658-G8100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-658-G8250 Supplier R&D Systems™ Supplier No. 11658G8250
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Measured by its ability to induce cytotoxicity using MPC-11 mouse B lymphocyte cells. The ED50 for this effect is 4.00-40.0 ng/mL.

Specifications

Accession Number O14793.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 2660
Molecular Weight (g/mol) MolecularWeight-observed: 10-14 kDa, under reducing conditions., MolecularWeight-theroretical: 12 kDa
Quantity 250 μg
Source Chinese Hamster Ovary cell line, CHO-derived GDF-8/Myostatin protein Asp267-Ser375
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Protein GDF-8/Myostatin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.