Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat NT-3, Animal-Free Protein

Catalog No. QK0580500
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QK0580500 500 μg
QK0580050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. QK0580500 Supplier R&D Systems™ Supplier No. QK0580500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human/Mouse/Rat/Porcine

No significant difference between EC50 of reference and test lots

Specifications

Accession Number P20783.1
For Use With (Application) Bioactivity
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 4908
Molecular Weight (g/mol) M.W. (Observed): Monomeric NT-3 protein only, M.W. (Theroretical): 27.3 kDa (dimer) & 13.7 kDa (monomer)
Quantity 500 μg
Source E. coli-derived NT-3 protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3
Conjugate Unconjugated
Reconstitution Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired.
Protein NT-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.