Learn More
Biomatik Corporation Recombinant Human Mucin-5B (MUC5B), partial
Supplier:  Biomatik Corporation RPC27157
Encompass_Preferred
Recombinant Human Mucin-5B (MUC5B), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MUC5B. Target Synonyms: MUC-5B; Cervical mucin; High molecular weight salivary mucin MG1; Mucin-5 subtype B, tracheobronchial; Sublingual gland mucin. Accession Number: Q9HC84. Expression Region: 4186~4295aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 18.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5751