Learn More
Biomatik Corporation Recombinant Human Mucosal addressin cell adhesion molecule 1 (MADCAM1), partial
Supplier:  Biomatik Corporation RPC26406
Encompass_Preferred
Recombinant Human Mucosal addressin cell adhesion molecule 1 (MADCAM1), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MADCAM1. Target Synonyms: MAdCAM-1hMAdCAM-1. Accession Number: Q13477. Expression Region: 19~317aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 34.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6356