Learn More
Biomatik Corporation Recombinant Human NACHT, LRR and PYD domains-containing protein 3 (NLRP3), partial
Supplier:  Biomatik Corporation RPC25978
Encompass_Preferred
Recombinant Human NACHT, LRR and PYD domains-containing protein 3 (NLRP3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NLRP3. Target Synonyms: Angiotensin/vasopressin receptor AII/AVP-like (CLR1.1; Cold-induced autoinflammatory syndrome 1 protein; Cryopyrin; PYRIN-containing APAF1-like protein 1; C1orf7; CIAS1; NALP3; PYPAF1; Caterpiller protein 1.1). Accession Number: Q96P20. Expression Region: 1~536aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 66.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4827