Learn More
Biomatik Corporation Recombinant Human NAD (+) hydrolase SARM1 (SARM1)
Supplier:  Biomatik Corporation RPC26393
Encompass_Preferred
Recombinant Human NAD (+) hydrolase SARM1 (SARM1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SARM1. Target Synonyms: Sterile alpha and Armadillo repeat protein (Sterile alpha motif domain-containing protein 2; MyD88-5; SAM domain-containing protein 2; Tir-1 homolog; KIAA0524; SAMD2; SARM). Accession Number: Q6SZW1. Expression Region: 28~724aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 80.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7929