Learn More
Abcam Recombinant Human NADPH oxidase 4 protein, 100UG
Supplier:  Abcam AB289740100UG
Encompass_Preferred
Recombinant Human NADPH oxidase 4 protein is a Human Full Length protein, in the 1 to 578 aa range, expressed in Cell free, with >=85% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: RENOX, NOX4, NADPH oxidase 4, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, Renal NAD(P)H-oxidase, KOX-1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2161873
Add to Cart
Request a Quote